Learn More
Cole-Parmer Cole-Parmer 1" NPT(F) T-fitting with 3/4" NPT(F), PVDF adapter installed in branch
Catalog No. 50633436 Shop All Cole-Parmer Products
Encompass_Preferred
This has a one inch NPT(F) connection on all three sides. The top has a 3/4 inch NPT(F) for fitting - reducing bushing installed.The self-cleaning electrode in 27003 series ranges from -5 to 110 degrees Celcius.
Catalog No. 50-633-436 Supplier Cole-Parmer Supplier No. 2700382
This has a one inch NPT(F) connection on all three sides. The top has a 3/4 inch NPT(F) for fitting - reducing bushing installed.The self-cleaning electrode in 27003 series ranges from -5 to 110 degrees Celcius.