Learn More
Grainger Tee Forged Steel 1 1/2 in Pipe Size FNPT
Catalog No. 502750608
Encompass_Preferred
The Tee Forged Steel 1-1/2 in Pipe Size FNPT is a forged steel tee fitting with female NPT threads. It is made from A105 forged carbon steel and has a maximum pressure rating of 2000 psi at 100°F. This tee fitting has a 1-1/2 inch pipe size opening and is designed for use in high pressure steam, water, oil and gas systems. The female NPT threads provide a tapered thread seal when used with pipe thread sealant. This durable fitting is ideal for creating a branch connection in piping systems.
Catalog No. 50-275-0608 Supplier Grainger Supplier No. 14J737
The Tee Forged Steel 1-1/2 in Pipe Size FNPT is a forged steel tee fitting with female NPT threads. It is made from A105 forged carbon steel and has a maximum pressure rating of 2000 psi at 100°F. This tee fitting has a 1-1/2 inch pipe size opening and is designed for use in high pressure steam, water, oil and gas systems. The female NPT threads provide a tapered thread seal when used with pipe thread sealant. This durable fitting is ideal for creating a branch connection in piping systems.