Learn More
Grainger Tee Forged Steel 2 in Pipe Size FNPT
Catalog No. 502750609
Encompass_Preferred
The Grainger Approved Tee Forged Steel 2" Pipe Size FNPT is a forged steel tee fitting with 2" female NPT threaded connections. It is made in the USA out of A105 forged carbon steel and is rated for a maximum pressure of 2000 psi at 650°F. This tee fitting allows you to branch off a 2" pipe line and is commonly used in steam, water, oil and natural gas applications. The female NPT threads provide a tapered thread seal when used with pipe thread sealant. This is an economical fitting for low pressure applications.
Catalog No. 50-275-0609 Supplier Grainger Supplier No. 14J738
The Grainger Approved Tee Forged Steel 2" Pipe Size FNPT is a forged steel tee fitting with 2" female NPT threaded connections. It is made in the USA out of A105 forged carbon steel and is rated for a maximum pressure of 2000 psi at 650°F. This tee fitting allows you to branch off a 2" pipe line and is commonly used in steam, water, oil and natural gas applications. The female NPT threads provide a tapered thread seal when used with pipe thread sealant. This is an economical fitting for low pressure applications.