Learn More
IDEX Health & Science Upchurch TEE HIGH PRESSURE MIXING
Catalog No. NC1919409
Encompass_Preferred
Idex Multi-Port High Pressure Connectors, Tee, Diameter Inner: 0.02 in., For Use With: 0.625 in. O.D. Tubing, Pressure Rating: 5000 psi, Swept Volume: 2.2 uL
Catalog No. NC1919409 Supplier IDEX Health & Science Upchurch Supplier No. U466
Idex Multi-Port High Pressure Connectors, Tee, Diameter Inner: 0.02 in., For Use With: 0.625 in. O.D. Tubing, Pressure Rating: 5000 psi, Swept Volume: 2.2 uL