Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Tee, Hose Barb Union, 5/8"" ID; 10/PK
Catalog No. 50630299
Encompass
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Catalog No. 50-630-299 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3061325
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).