Learn More
Heidolph™ Tee for Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
SDP
Catalog No. 13900927 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-927 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-927 Supplier Heidolph™ Supplier No. 036251830
Add to Cart
Add to Cart
Tee connects condenser and distributor.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware set A, A1, A1C, A2, A2C, and AC
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov