Learn More
IDEX Health & Science Upchurch TEE PEEK .020 THRU HOLE
Catalog No. NC1919102
Encompass_Preferred
Idex Low-Pressure PEEK Tees, Diameter Inner: 0.50 mm, 0.020 in., Tees and Crosses, Pressure Rating: 1000 psi
Catalog No. NC1919102 Supplier IDEX Health & Science Upchurch Supplier No. P712
Idex Low-Pressure PEEK Tees, Diameter Inner: 0.50 mm, 0.020 in., Tees and Crosses, Pressure Rating: 1000 psi