Learn More
IDEX Health & Science Upchurch TEE PEEK .050 THRU HOLE
Catalog No. NC1919103
Encompass_Preferred
Idex Low-Pressure PEEK Tees, Diameter Inner: 1.25 mm, 0.050 in., Union, Pressure Rating: 1000 psi
Catalog No. NC1919103 Supplier IDEX Health & Science Upchurch Supplier No. P713
Idex Low-Pressure PEEK Tees, Diameter Inner: 1.25 mm, 0.050 in., Union, Pressure Rating: 1000 psi