Learn More
IDEX Health & Science Upchurch TEE PRESSURE GAUGE
Catalog No. NC1919400
Encompass_Preferred
Idex High-Pressure Tees, Diameter Outer: 1.02 mm, Stainless Steel, Diameter Inner: 0.04 in., For Use With: 1/16 in. O.D. Tubing
Catalog No. NC1919400 Supplier IDEX Health & Science Upchurch Supplier No. U433
Idex High-Pressure Tees, Diameter Outer: 1.02 mm, Stainless Steel, Diameter Inner: 0.04 in., For Use With: 1/16 in. O.D. Tubing