Learn More
IDEX Health & Science Upchurch TEE TEFZEL .020 THRU HOLE
Catalog No. NC1919093
Encompass_Preferred
Idex Tefzel Low-Pressure Unions, Diameter Inner: 0.05 in., Natural, Tee, ETFE, Pressure Rating: 1000 psi, Swept Volume: 2.9 uL
Catalog No. NC1919093 Supplier IDEX Health & Science Upchurch Supplier No. P632
Idex Tefzel Low-Pressure Unions, Diameter Inner: 0.05 in., Natural, Tee, ETFE, Pressure Rating: 1000 psi, Swept Volume: 2.9 uL