Learn More
IDEX Health & Science Upchurch TEE TEFZEL .050 THRU HOLE
Supplier:  IDEX Health & Science Upchurch P633
Encompass_Preferred
Idex Tefzel Low-Pressure Unions, Diameter Inner: 0.05 in., Natural, Tee, ETFE, Pressure Rating: 1000 psi, Swept Volume: 17.5 uL
Catalog No. NC1919094