Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin Compression union tee, PFA, 1/8""
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3132029
Encompass
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. 50-631-993