Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TeenPrep™ Lysing Matrix J

Catalog No. MP116936050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 x 15 mL
25 x 15 mL
50 x 15 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116936050 50 x 15 mL
MP116936025 25 x 15 mL
MP116936005 5 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP116936050 Supplier MP Biomedicals™ Supplier No. 116936050
Add to Cart
Add to Cart

TeenPrep™ Lysing Matrix J, 15 mL Tube contains 2 mm yellow zirconia beads and 1.6 mm aluminum oxide particles. Will not bind nucleic acids.

  • Tube contains 2 mm yellow zirconia beads and 1.6 mm aluminum oxide particles.
  • Used to grind animal and plant tissues, fixed samples and lyse fungi.
  • For use on FastPrep™ instruments or any bead beater.

Certifications

Certified RNase and DNase free.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 15 mL
Sample Type Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Fixed Samples, Coral Emulsions, Mold, Animal Tissues, Fungi, Plants, Fixed Samples, Coral Emulsions, Mold
DNase DNase Free
For Use With (Equipment) FastPrep-24, FastPrep 120; FastPrep-96
For Use With (Application) Sample Preparation, Sample lysis, Nucleic Acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis, Gene function, Gene regulation
Product Type Lysing Matrix J
Quantity 50 x 15 mL
RNase RNase Free
Sample Volume Up to 2 g per prep

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.