Learn More
Research Products International Corp Teeny Tough-Tag for Laser, White, 0.91 x 0.32 Inches (28 mm x 9 mm), 3850 per Package
SDP
Supplier:  Research Products International Corp 247151
Encompass
Teeny Tough-Tags are pre-cut labels for 0.2 ml tubes. Ideal size allows perfect fit around 0.2ml cycling tubes. Unique labels are thermocycler compatible and narrow size allows exact placement on upper wall of micro-tubes, above the heating block. Tags are designed to withstand boiling water baths, freezer and cryogenic temperatures, organic solvents, and caustic agents.
Catalog No. 50-550-259