Learn More
Crescent Chemical Co Inc TEEPOL 610 PRACT.
SDP
Supplier:  Crescent Chemical Co Inc SE35796.01
Encompass_Preferred
Teepol 610 pract. 32 % aqueous solution of sodium C9-C11 alkylethersulfates.A dilution of 1:20 has pH 7.6. For determination of iron in serum (1). Teepol 710 originally used by the author is no longer available. A modification of Lauber's procedure can be su
Catalog No. 50-248-178