Learn More
AVANTOR MASTERFLEX (VWR LLC) TEES 1/8FTFT X 3/8FTFT 10/PK
Catalog No. 502742338
Encompass
Masterflex Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/8IN ID x 3/8IN ID x 1/8IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2338 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061073
Masterflex Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/8IN ID x 3/8IN ID x 1/8IN NPT(M)10/PK, Avantor