Learn More
AVANTOR MASTERFLEX (VWR LLC) TEES 3/4IN X 3/4IN 5/PK
Catalog No. 502742326
Encompass
Masterflex Fitting, Natural Polypropylene, Tee, Hose Barb to Thread Adapter, 3/4IN ID x 3/4IN ID x 3/4IN NPT(M)5/PK, Avantor
Catalog No. 50-274-2326 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061087
Masterflex Fitting, Natural Polypropylene, Tee, Hose Barb to Thread Adapter, 3/4IN ID x 3/4IN ID x 3/4IN NPT(M)5/PK, Avantor