Learn More
AVANTOR MASTERFLEX (VWR LLC) TEES 3/8FTFT X 1/4FTFT 10/PK
Catalog No. 502742334
Encompass
Masterflex Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 1/4IN ID x 1/4IN ID x 3/8IN NPT(M)10/PK, Avantor
Catalog No. 50-274-2334 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4061077
Masterflex Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 1/4IN ID x 1/4IN ID x 3/8IN NPT(M)10/PK, Avantor