Learn More
Analytical Sales & Services Teflon Adapter, 1/4-28 Female to 1/4-28 Male
Catalog No. 5000804659
Encompass
Teflon Adapter 0.040" ID 1/4-28 Female to 1/4-28 Male
Qty/Pkg: EACH
Catalog No. 50-008-04659 Supplier Analytical Sales & Services Supplier No. 59671
Add to Cart
Add to Cart
Teflon Adapter 0.040" ID 1/4-28 Female to 1/4-28 Male
Qty/Pkg: EACH