Learn More
VWR INTERNATIONAL Chemware Beakers, Teflon PFA, Saint-Gobain Performance Plastics, 125 mL
Catalog No. NC1778600
Encompass
These low metal ion content PFA beakers are ideal for trace metal analysis and ultra-pure applications.
Catalog No. NC1778600 Supplier VWR INTERNATIONAL Supplier No. 13917582
Add to Cart
Add to Cart
These low metal ion content PFA beakers are ideal for trace metal analysis and ultra-pure applications.