Learn More
Waters Corp Waters Corporation Tubing, PTFE, 0.062 in. × 0.01 in. × 0.75 in., 5/pk
Catalog No. 50465952 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PTFE tubing is designed for various laboratory applications. It features precise dimensions and comes in a convenient pack of five.
  • Made from PTFE for chemical inertness and high temperature resistance
  • Outer diameter of 0.062 in.
  • Inner diameter of 0.01 in.
  • Length of 0.75 in.
  • Supplied in a pack of five
  • Suitable as a spare part for various systems
Catalog No. 50-465-952 Supplier Waters Corp Supplier No. 186002690
This PTFE tubing is designed for various laboratory applications. It features precise dimensions and comes in a convenient pack of five.
  • Made from PTFE for chemical inertness and high temperature resistance
  • Outer diameter of 0.062 in.
  • Inner diameter of 0.01 in.
  • Length of 0.75 in.
  • Supplied in a pack of five
  • Suitable as a spare part for various systems