Learn More
J Kem Scientific Inc TEFLON COATED NEEDLE THERMOCOU
SDP
Catalog No. NC9957151
Encompass
Needles are sealed on the end for liquid tight applications. Available in 3.50 and 70 lengths. Stainless steel. Also available without PTFE coating. Each comes with 72 inches of attached PTFE extension wire.
Catalog No. NC9957151 Supplier J Kem Scientific Inc Supplier No. HN7TTEF
Needles are sealed on the end for liquid tight applications. Available in 3.50 and 70 lengths. Stainless steel. Also available without PTFE coating. Each comes with 72 inches of attached PTFE extension wire.