Learn More
Cole-Parmer PTFE Coated Lab Spatula w/ 1.5" Flat Taper x 1.25" Flat Round Ends; 1/Pk
Catalog No. NC9979753 Shop All Cole-Parmer Products
Encompass_Preferred
Double-ended spatulas are completely covered with extra heavy-duty coating of PTFE—prevents buildup on surfaces or loss of critical quantities due to sticking. Will not scratch or mar delicate surfaces.
Catalog No. NC9979753 Supplier Cole-Parmer Supplier No. 0636911
Double-ended spatulas are completely covered with extra heavy-duty coating of PTFE—prevents buildup on surfaces or loss of critical quantities due to sticking. Will not scratch or mar delicate surfaces.