Learn More
Fuel Cell Earth Teflon PTFE DISP 30, Fluoropolymer Dispersion, 250mL
Catalog No. 501090481
Encompass_Preferred
TEFLON PTFE by DuPont (Polytetrafluoroethylene) is known for its exceptional resistance to high temperatures and chemical, corrosion, and stress crack resistance. The mechanical toughness, electrical, and low-friction properties of Teflon make it the preferred plastic for a host of applications and different processing techniques. DISP 30 aqueous dispersion provides: Inertness to nearly all industrial chemicals and solvents, Stability at high temperatures, Excellent dielectric properties, Lowest coefficient of friction of any solid material, Excellent weatherability, Non-stick characteristics.
Catalog No. 50-109-0481 Supplier Fuel Cell Earth Supplier No. DISP30250
TEFLON PTFE by DuPont (Polytetrafluoroethylene) is known for its exceptional resistance to high temperatures and chemical, corrosion, and stress crack resistance. The mechanical toughness, electrical, and low-friction properties of Teflon make it the preferred plastic for a host of applications and different processing techniques. DISP 30 aqueous dispersion provides: Inertness to nearly all industrial chemicals and solvents, Stability at high temperatures, Excellent dielectric properties, Lowest coefficient of friction of any solid material, Excellent weatherability, Non-stick characteristics.