Learn More
Chemglass Life Sciences PTFE Envelope Gasket For 300mm Duran Flange, 11-5/8" ID X 15" OD, 1/8" Silicone Core
SDP
Catalog No. 50872163
Encompass_Preferred
This envelope style gasket is machined using chemically resistant virgin PTFE and has a silicone core. The larger sealing surface results in improved sealing under full vacuum, as opposed to other o-ring materials. Gasket for use with 300mm Duran Flange on 30L and larger reactors.
Catalog No. 50-872-163 Supplier Chemglass Life Sciences Supplier No. CG1968G01
This envelope style gasket is machined using chemically resistant virgin PTFE and has a silicone core. The larger sealing surface results in improved sealing under full vacuum, as opposed to other o-ring materials. Gasket for use with 300mm Duran Flange on 30L and larger reactors.