Learn More
Chemglass Life Sciences 30mm PTFE Flake Retaining Cup
SDP
Catalog No. 50997772
Encompass_Preferred
30mm PTFE Flake Retaining Cup. Cup is designed to catch all flaking that occurs with PTFE stirrer bearings. Made entirely of PTFE, the complete item consists of a compression fitting, deep perforated PTFE cup and a 50 micron porosity PTFE filter disc.
Catalog No. 50-997-772 Supplier Chemglass Life Sciences Supplier No. CG209322
30mm PTFE Flake Retaining Cup. Cup is designed to catch all flaking that occurs with PTFE stirrer bearings. Made entirely of PTFE, the complete item consists of a compression fitting, deep perforated PTFE cup and a 50 micron porosity PTFE filter disc.