Learn More
Analytical Sales & Services Teflon Piston Seals for Agilent 1050, 1100, 1200, 1260
Catalog No. 5000806970
Encompass
Agilent 1050 1100 1200 1260 1290 Analytical Pumps Replacement Teflon Piston Seals OEM# 5062-85167 5063-6589
Qty/Pkg: 10PK
Catalog No. 50-008-06970 Supplier Analytical Sales & Services Supplier No. 930095
Agilent 1050 1100 1200 1260 1290 Analytical Pumps Replacement Teflon Piston Seals OEM# 5062-85167 5063-6589
Qty/Pkg: 10PK