Learn More
Ace Glass, Inc. TEFLON RETAINER RING
SDP
Catalog No. NC0094095
Encompass_Preferred
PTFE retaining ring used with chromatography column adapters. The adapters are used as a chromatography column end fitting that allows for the connection of #80 Ace-Thred joints to tubing connectors with NPT threads.
Catalog No. NC0094095 Supplier Ace Glass, Inc. Supplier No. 585752
PTFE retaining ring used with chromatography column adapters. The adapters are used as a chromatography column end fitting that allows for the connection of #80 Ace-Thred joints to tubing connectors with NPT threads.