Learn More
Fritsch Milling And Sizing Inc Teflon Seal Ring for Grinding Bowls 80 Ml
SDP
Catalog No. 501534569
Encompass
Replacement seal ring made of PTFE, diameter 80/65 mm
for all grinding bowls classic line with 80 ml volume (except agate).
Catalog No. 50-153-4569 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.4230.20
Replacement seal ring made of PTFE, diameter 80/65 mm
for all grinding bowls classic line with 80 ml volume (except agate).