Learn More
Analytical Sales & Services Teflon® Tubing, 1/8" OD X 1/16" ID X 10'
Catalog No. 50453706
Encompass
This item has a minimum order quantity of 2 per supplier requirements. PTFE TUBING 1/8 X 1/16ID 10' Maximum Pressure: 500psi
Catalog No. 50-453-706 Supplier Analytical Sales & Services Supplier No. 415234
This item has a minimum order quantity of 2 per supplier requirements. PTFE TUBING 1/8 X 1/16ID 10' Maximum Pressure: 500psi