Learn More
Analytical Sales & Services Tefzel Flangeless Male Nut for 1/8" OD Tubing
Catalog No. 5000806883
Encompass
Tefzel Flangeless Male Nut for 1/8" OD Tubing 1/4-28
Qty/Pkg: 10Pk
Catalog No. 50-008-06883 Supplier Analytical Sales & Services Supplier No. 59345X
Tefzel Flangeless Male Nut for 1/8" OD Tubing 1/4-28
Qty/Pkg: 10Pk