Learn More
Analytical Sales & Services Tefzel Large Bore Flangeless Ferrule for 3/16'' OD Tubing
Catalog No. 5000804542
Encompass
Tefzel Large Bore Flangeless Ferrule\n\n• For 3/16" OD Tubing\n• Color: Blue\n\nQty/Pkg: 10
Catalog No. 50-008-04542 Supplier Analytical Sales & Services Supplier No. 59133X
Tefzel Large Bore Flangeless Ferrule\n\n• For 3/16" OD Tubing\n• Color: Blue\n\nQty/Pkg: 10