Learn More
Perkin Elmer US LLC Tefzel Nut for Reverse Ferrule, 0.125 in
Catalog No. 502112420
Encompass_Preferred
Included in the HPLC Starter Kit (N2930802). Maximum Pressure: 400 psi Tubing Size: 1/16 inch O.D. Thread: 1/4-28
Catalog No. 50-211-2420 Supplier Perkin Elmer US LLC Supplier No. N2601189
Included in the HPLC Starter Kit (N2930802). Maximum Pressure: 400 psi Tubing Size: 1/16 inch O.D. Thread: 1/4-28