Learn More
Bio Rad Laboratories TEFZEL PLUG
Catalog No. 509011191
Encompass
Plug, Ports; Bio-Rad; Tefzel; For plugging unused ports on valves, columns; 1/4 to 28 male connection; Qty: 5
Catalog No. 50-901-1191 Supplier Bio Rad Laboratories Supplier No. 7500563
Plug, Ports; Bio-Rad; Tefzel; For plugging unused ports on valves, columns; 1/4 to 28 male connection; Qty: 5