Learn More
Analytical Sales & Services Tefzel Small Valve Ferrule for 1/16" OD Tubing
Catalog No. 5000804599
Encompass
Tefzel Small Valve Flangeless Ferrule for 1/16" OD Tubing Natural
Qty/Pkg: 10PK
Catalog No. 50-008-04599 Supplier Analytical Sales & Services Supplier No. 59240X
Tefzel Small Valve Flangeless Ferrule for 1/16" OD Tubing Natural
Qty/Pkg: 10PK