Learn More
IDEX Health & Science Upchurch TEFZEL TUBING .030X.062X100'
Catalog No. NC1919419
Encompass_Preferred
Idex ETFE Chromatography Tubing, Natural, Diameter Inner: 0.03 in., Overall Length: 100 ft., Pressure: 500 to 4000 psi
Catalog No. NC1919419 Supplier IDEX Health & Science Upchurch Supplier No. 1528XL
Idex ETFE Chromatography Tubing, Natural, Diameter Inner: 0.03 in., Overall Length: 100 ft., Pressure: 500 to 4000 psi