Learn More
IDEX Health & Science Upchurch TEFZEL TUBING .030X.062X50
Catalog No. NC1919421
Encompass_Preferred
Idex ETFE Chromatography Tubing, Natural, Diameter Inner: 0.03 in., Overall Length: 50 ft., Pressure: 500 to 4000 psi
Catalog No. NC1919421 Supplier IDEX Health & Science Upchurch Supplier No. 1528L
Idex ETFE Chromatography Tubing, Natural, Diameter Inner: 0.03 in., Overall Length: 50 ft., Pressure: 500 to 4000 psi