Learn More
IDEX Health & Science Upchurch TEFZEL TUBING .062X.125X100'
Catalog No. NC1919418
Encompass_Preferred
Idex ETFE Chromatography Tubing, Natural, Diameter Inner: 0.055 to 0.065 in., Overall Length: 100 ft., Pressure: 1000 psi
Catalog No. NC1919418 Supplier IDEX Health & Science Upchurch Supplier No. 1530XL
Idex ETFE Chromatography Tubing, Natural, Diameter Inner: 0.055 to 0.065 in., Overall Length: 100 ft., Pressure: 1000 psi