Learn More
Analytical Sales & Services Tefzel ZDV Union Assembly for 1/16" Tubing
Catalog No. 5000804657
Encompass
Tefzel ZDV Union Assembly for 1/16" Tubing 1/4-28 Threads
Qty/Pkg: EACH
Catalog No. 50-008-04657 Supplier Analytical Sales & Services Supplier No. 59630
Tefzel ZDV Union Assembly for 1/16" Tubing 1/4-28 Threads
Qty/Pkg: EACH