Learn More
Med Vet International Transparent Film Dressing With Pad 3M Tegaderm +Pad 6 X 6 inch Frame Style DelIVery Square Sterile, 3588, Each
Catalog No. 502831385
Encompass_Preferred
Tegaderm +Pad film dressing with nonadherent pad is a composite dressing A sterile, waterproof bacterial barrier consists of a non-adherent absorbent pad bonded to a larger thin film transparent dressing Breathable film allows skin to function normally with good exchange of moisture vapor and oxygen Moist wound environment enhances healing and lessens pain Nonadherent pad won't stick to the wound, allowing normal wound healing processes to occur with less pain and trauma
Catalog No. 50-283-1385 Supplier Med Vet International Supplier No. 360349EA
Tegaderm +Pad film dressing with nonadherent pad is a composite dressing A sterile, waterproof bacterial barrier consists of a non-adherent absorbent pad bonded to a larger thin film transparent dressing Breathable film allows skin to function normally with good exchange of moisture vapor and oxygen Moist wound environment enhances healing and lessens pain Nonadherent pad won't stick to the wound, allowing normal wound healing processes to occur with less pain and trauma