Learn More
Medchemexpress LLC Tegafur-Uracil | 74578-38-4 | 99.77% | 312.25 | 5 MG
SDP
Catalog No. 5000395257
Encompass_Preferred
Tegafur-Uracil (UFT; BMS-200604) is an effective oral chemotherapy drug based on fluoropyrimidine. It inhibits thymidylate synthase and can be used in combination with leucovorin (LV) and polysaccharide K (PSK) for research on solid tumors.
  • Effective oral chemotherapy drug based on fluoropyrimidine.
  • Inhibits thymidylate synthase.
  • Can be used with leucovorin (LV) and polysaccharide K (PSK) for solid tumor research.
  • For research use only.
Catalog No. 50-003-95257 Supplier Medchemexpress LLC Supplier No. HY161105MG
Tegafur-Uracil (UFT; BMS-200604) is an effective oral chemotherapy drug based on fluoropyrimidine. It inhibits thymidylate synthase and can be used in combination with leucovorin (LV) and polysaccharide K (PSK) for research on solid tumors.
  • Effective oral chemotherapy drug based on fluoropyrimidine.
  • Inhibits thymidylate synthase.
  • Can be used with leucovorin (LV) and polysaccharide K (PSK) for solid tumor research.
  • For research use only.