Learn More
Medchemexpress LLC Tegafur-Uracil | 74578-38-4 | 99.8% | 312.25 | 25 MG
SDP
Catalog No. 5000395387
Encompass_Preferred
Tegafur-Uracil (UFT; BMS-200604) is an effective oral chemotherapy drug based on fluoropyrimidine. It inhibits thymidylate synthase and can be used in combination with leucovorin (LV) and polysaccharide K (PSK) for research on solid tumors.
  • Inhibits thymidylate synthase.
  • Effective oral chemotherapy drug.
  • Can be used in combination with leucovorin (LV) and polysaccharide K (PSK) for research on solid tumors.
  • Enhances anti-tumor activity in mice when combined with folinic acid and protein-bound polysaccharides.
Catalog No. 50-003-95387 Supplier Medchemexpress LLC Supplier No. 25MGHY16110
Tegafur-Uracil (UFT; BMS-200604) is an effective oral chemotherapy drug based on fluoropyrimidine. It inhibits thymidylate synthase and can be used in combination with leucovorin (LV) and polysaccharide K (PSK) for research on solid tumors.
  • Inhibits thymidylate synthase.
  • Effective oral chemotherapy drug.
  • Can be used in combination with leucovorin (LV) and polysaccharide K (PSK) for research on solid tumors.
  • Enhances anti-tumor activity in mice when combined with folinic acid and protein-bound polysaccharides.