Learn More
Medchemexpress LLC Tegaserod (maleate) | 189188-57-6 | 99.9% | C20H27N5O5 | 1 G
SDP
Catalog No. 5000353760
Encompass_Preferred
Tegaserod maleate is an orally active compound that functions as a serotonin receptor 4 agonist and a serotonin receptor 2B antagonist. It demonstrates inhibitory activity against 5-HT2A, 5-HT2B, and 5-HT2C receptors. This compound has been observed to induce apoptosis in tumor cells, inhibit PI3K/Akt/mTOR signaling, and reduce S6 phosphorylation. It shows promise for anti-tumor applications and research into irritable bowel syndrome.
  • Orally active serotonin receptor 4 agonist.
  • Serotonin receptor 2B antagonist.
  • Induces tumor cell apoptosis.
  • Inhibits PI3K/Akt/mTOR signaling and decreases S6 phosphorylation.
  • Exhibits anti-tumor activity, delays tumor growth, reduces metastases, and increases survival.
  • Potential for irritable bowel syndrome research.
Catalog No. 50-003-53760 Supplier Medchemexpress LLC Supplier No. HY14153A1G
Tegaserod maleate is an orally active compound that functions as a serotonin receptor 4 agonist and a serotonin receptor 2B antagonist. It demonstrates inhibitory activity against 5-HT2A, 5-HT2B, and 5-HT2C receptors. This compound has been observed to induce apoptosis in tumor cells, inhibit PI3K/Akt/mTOR signaling, and reduce S6 phosphorylation. It shows promise for anti-tumor applications and research into irritable bowel syndrome.
  • Orally active serotonin receptor 4 agonist.
  • Serotonin receptor 2B antagonist.
  • Induces tumor cell apoptosis.
  • Inhibits PI3K/Akt/mTOR signaling and decreases S6 phosphorylation.
  • Exhibits anti-tumor activity, delays tumor growth, reduces metastases, and increases survival.
  • Potential for irritable bowel syndrome research.