Learn More
Medchemexpress LLC Tegaserod (maleate) | 189188-57-6 | 99.9% | C20H27N5O5 | 1 ML
SDP
Catalog No. 5000356710
Encompass_Preferred
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4) agonist and a 5-HT2B receptor antagonist. It demonstrates anti-tumor activity by inducing tumor cell apoptosis and blunting PI3K/Akt/mTOR signaling. It is also being researched for its potential in treating irritable bowel syndrome (IBS) and diabetic gastropathy.
  • Potent agonist of 5-HT4 receptor
  • Antagonist of 5-HT2B receptor
  • Induces tumor cell apoptosis
  • Blunts PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Accelerates gastric emptying rate
Catalog No. 50-003-56710 Supplier Medchemexpress LLC Supplier No. HY14153A10MM/1ML
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4) agonist and a 5-HT2B receptor antagonist. It demonstrates anti-tumor activity by inducing tumor cell apoptosis and blunting PI3K/Akt/mTOR signaling. It is also being researched for its potential in treating irritable bowel syndrome (IBS) and diabetic gastropathy.
  • Potent agonist of 5-HT4 receptor
  • Antagonist of 5-HT2B receptor
  • Induces tumor cell apoptosis
  • Blunts PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Accelerates gastric emptying rate