Learn More
Medchemexpress LLC Tegaserod (maleate) | 189188-57-6 | 99.9% | C20H27N5O5 | 100 MG
SDP
Catalog No. 5000353764
Encompass_Preferred
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist and a 5-HT2B receptor antagonist. It causes tumor cell apoptosis, blunts PI3K/Akt/mTOR signaling, and decreases S6 phosphorylation. This compound exhibits anti-tumor activity and shows potential for irritable bowel syndrome (IBS) research.
  • Orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist
  • 5-HT2B receptor antagonist
  • Causes tumor cell apoptosis
  • Blunts PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Has anti-tumor activity
  • Potential for irritable bowel syndrome (IBS) research
Catalog No. 50-003-53764 Supplier Medchemexpress LLC Supplier No. HY14153A100MG
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist and a 5-HT2B receptor antagonist. It causes tumor cell apoptosis, blunts PI3K/Akt/mTOR signaling, and decreases S6 phosphorylation. This compound exhibits anti-tumor activity and shows potential for irritable bowel syndrome (IBS) research.
  • Orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist
  • 5-HT2B receptor antagonist
  • Causes tumor cell apoptosis
  • Blunts PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Has anti-tumor activity
  • Potential for irritable bowel syndrome (IBS) research