Learn More
Medchemexpress LLC Tegaserod (maleate) | 189188-57-6 | 99.9% | C20H27N5O5 | 25 MG
SDP
Catalog No. 5000352793
Encompass_Preferred
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist and a 5-HT2B receptor antagonist. It induces tumor cell apoptosis, blunts PI3K/Akt/mTOR signaling, and decreases S6 phosphorylation, exhibiting anti-tumor activity.
  • Orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist
  • 5-HT2B receptor antagonist
  • Induces tumor cell apoptosis
  • Blunts PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Exhibits anti-tumor activity
  • Potential for irritable bowel syndrome (IBS) research
Catalog No. 50-003-52793 Supplier Medchemexpress LLC Supplier No. 25MGHY14153A
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist and a 5-HT2B receptor antagonist. It induces tumor cell apoptosis, blunts PI3K/Akt/mTOR signaling, and decreases S6 phosphorylation, exhibiting anti-tumor activity.
  • Orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist
  • 5-HT2B receptor antagonist
  • Induces tumor cell apoptosis
  • Blunts PI3K/Akt/mTOR signaling
  • Decreases S6 phosphorylation
  • Exhibits anti-tumor activity
  • Potential for irritable bowel syndrome (IBS) research