Learn More
Medchemexpress LLC Tegaserod maleate | 189188-57-6 | 99.9% | C20H27N5O5 | 50 MG
SDP
Catalog No. 5000356708
Encompass_Preferred
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist and a 5-HT2B receptor antagonist. This compound induces tumor cell apoptosis, attenuates PI3K/Akt/mTOR signaling, and reduces S6 phosphorylation. It demonstrates anti-tumor activity and is being investigated for its potential in irritable bowel syndrome (IBS) research.
  • Orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist
  • 5-HT2B receptor antagonist
  • Induces tumor cell apoptosis
  • Attenuates PI3K/Akt/mTOR signaling
  • Reduces S6 phosphorylation
  • Demonstrates anti-tumor activity
  • Potential for irritable bowel syndrome (IBS) research
  • For research use only
Catalog No. 50-003-56708 Supplier Medchemexpress LLC Supplier No. HY14153A50MG
Tegaserod maleate is an orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist and a 5-HT2B receptor antagonist. This compound induces tumor cell apoptosis, attenuates PI3K/Akt/mTOR signaling, and reduces S6 phosphorylation. It demonstrates anti-tumor activity and is being investigated for its potential in irritable bowel syndrome (IBS) research.
  • Orally active serotonin receptor 4 (HTR4; 5-HT4R) agonist
  • 5-HT2B receptor antagonist
  • Induces tumor cell apoptosis
  • Attenuates PI3K/Akt/mTOR signaling
  • Reduces S6 phosphorylation
  • Demonstrates anti-tumor activity
  • Potential for irritable bowel syndrome (IBS) research
  • For research use only