Learn More
Selleck Chemical LLC Tegaserod Maleate 25mg 189188-57-6
SDP
Supplier:  Selleck Chemical LLC S540125MG
Encompass_Preferred
Tegaserod Maleate is a hydrogen maleate salt form of tegaserod, which is a 5-HT4 receptor partial agonist and binds with high affinity to 5-HT4 receptors. It has limited affinity for 5-HT1 receptors and no appreciable affinity for other 5-HT receptors, muscarinic, adrenergic, dopaminergic or opiate receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8376