Learn More
TARGETMOL CHEMICALS INC Tegaserod maleate 50MG
SDP
Catalog No. 502878285
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Tegaserod maleate (SDZ-HTF-919) is a 5-HT4 agonist manufactured by Novartis for managing irritable bowel syndrome and constipation. It was the only drug approved by the United States Food and Drug Administration for relieving abdominal discomfort, bloating, and constipation associated with irritable bowel syndrome. On March 30, 2007, the FDA requested that Novartis withdraw Zelnorm from the market due to alleged increased risks of heart attack or stroke associated with its use. Purity 99.06%
Catalog No. 50-287-8285 Supplier TARGETMOL CHEMICALS INC Supplier No. T155150MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Tegaserod maleate (SDZ-HTF-919) is a 5-HT4 agonist manufactured by Novartis for managing irritable bowel syndrome and constipation. It was the only drug approved by the United States Food and Drug Administration for relieving abdominal discomfort, bloating, and constipation associated with irritable bowel syndrome. On March 30, 2007, the FDA requested that Novartis withdraw Zelnorm from the market due to alleged increased risks of heart attack or stroke associated with its use. Purity 99.06%