Learn More
Selleck Chemical LLC Tegaserod Maleate S5401-1g
SDP
Catalog No. 503110214
Encompass_Preferred
Tegaserod Maleate(HTF-919) is a hydrogen maleate salt form of tegaserod which is a 5-HT4 receptor partial agonist and binds with high affinity to 5-HT4 receptors It has limited affinity for 5-HT1 receptors and no appreciable affinity for other 5-HT receptors muscarinic adrenergic dopaminergic or opiate receptors
Catalog No. 50-311-0214 Supplier Selleck Chemical LLC Supplier No. S54011G
Tegaserod Maleate(HTF-919) is a hydrogen maleate salt form of tegaserod which is a 5-HT4 receptor partial agonist and binds with high affinity to 5-HT4 receptors It has limited affinity for 5-HT1 receptors and no appreciable affinity for other 5-HT receptors muscarinic adrenergic dopaminergic or opiate receptors